Boc-Leu-OSu

Supplier: BACHEM AMERICAS INC

4001880.0005 4001880.0100
A-1860.0005BAEA 185.46 CAD
A-1860.0005BA A-1860.0100BA
Boc-Leu-OSu
Boc-Leu-OSu

Sequence: Boc-Leu-OSu


Bachem additionally offers deuterated and ¹⁵N-labeled leucine: Fmoc-[D₁₀]Leu-OH (B-4120) and Fmoc-[¹⁵N]Leu-OH (B-2655).


Used in an optimization study for obtaining enantiomerically pure thiodipeptides.

Formula: C₁₅H₂₄N₂O₆
Storage Temperature: Freezer
MDL Number: MFCD00037913
CAS Number: 3392-09-4
EINECS: 222-232-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR