N-(tert-Butoxycarbonyl)-D-leucine hydrate

Supplier: BACHEM AMERICAS INC

Synonyms: N-Boc-D-leucine, Boc-D-Leu-OH

4002976.0005 4002976.0025 4002976.0100
A-1855.0005BAEA 175.66 CAD
A-1855.0005BA A-1855.0025BA A-1855.0100BA
N-(tert-Butoxycarbonyl)-D-leucine hydrate
N-(tert-Butoxycarbonyl)-D-leucine hydrate

Sequence: Boc-D-Leu-OH · H₂O


Bachem additionally offers deuterated and ¹⁵N-labeled leucine: Fmoc-[D₁₀]Leu-OH (B-4120) and Fmoc-[¹⁵N]Leu-OH (B-2655).

Formula: C₁₁H₂₁NO₄
Storage Temperature: Ambient
MDL Number: MFCD00038294
CAS Number: 16937-99-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR