N-(tert-Butoxycarbonyl)-L-leucine hydrate
Supplier: BACHEM AMERICAS INC
Synonyms:
Boc-L-Leucine, BOC-L-Leucine hydrate, Boc-Leu-OH hydrate, N-Boc-L-leucine hydrate
4001021.0100
4001021.0500
4001021.1000
A-1850.0100BAEA
232.96
CAD
A-1850.0100BA
A-1850.0500BA
A-1850.1000BA
N-(tert-Butoxycarbonyl)-L-leucine hydrate
N-(tert-Butoxycarbonyl)-L-leucine hydrate
Sequence: Boc-Leu-OH · H₂O
Bachem additionally offers deuterated and ¹⁵N-labeled leucine: Fmoc-[D₁₀]Leu-OH (B-4120) and Fmoc-[¹⁵N]Leu-OH (B-2655).
Formula:
C₁₁H₂₁NO₄·xH₂O MW: 231.29 g/mol (anhy) Storage Temperature: Ambient |
MDL Number:
MFCD00066067 CAS Number: 13139-15-6 EINECS: 236-073-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...