Boc-D-isoleucine

Supplier: BACHEM AMERICAS INC

4000565.0001 4000565.0005
A-1830.0001BAEA 627.31 CAD
A-1830.0001BA A-1830.0005BA
Boc-D-isoleucine
Boc-D-isoleucine

Sequence: Boc-D-Ile-OH

Formula: C₁₁H₂₁NO₄
Storage Temperature: Ambient
MDL Number: MFCD00069989
CAS Number: 55721-65-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR