Boc-L-isoleucine
Supplier: BACHEM AMERICAS INC
4001026.0100
4001026.0500
A-1825.0100BAEA
256.51
CAD
A-1825.0100BA
A-1825.0500BA
Boc-L-isoleucine
Boc-L-isoleucine
Sequence: Boc-Ile-OH
Formula:
C₁₁H₂₁NO₄ Storage Temperature: Ambient |
MDL Number:
MFCD00038324 CAS Number: 13139-16-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...