N(α)-Boc-L-histidine
Supplier: BACHEM AMERICAS INC
Synonyms:
Boc-His-OH
4000667.0005
4000667.0025
4000667.0100
A-1765.0005BAEA
180.91
CAD
A-1765.0005BA
A-1765.0025BA
A-1765.0100BA
N(α)-Boc-L-histidine
N(α)-Boc-L-histidine
Sequence: Boc-His-OH
The atoms of the imidazole ring of histidine can be numbered in two different ways. Bachem as well as organic chemists designate the position of N-τ as 1 and the position of N-π as 3, whereas biochemists generally number N-π, the nitrogen atom adjacent to the side chain, as 1, and N-τ as 3.
Formula:
C₁₁H₁₇N₃O₄ MW: 255.3 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD00065576 CAS Number: 17791-52-5 EINECS: 241-768-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...