Boc-glycine-N-hydroxysuccinimide ester

Supplier: BACHEM AMERICAS INC

4001333.0005 4001333.0025
A-1735.0005BAEA 187.69 CAD
A-1735.0005BA A-1735.0025BA
Boc-glycine-N-hydroxysuccinimide ester
Boc-glycine-N-hydroxysuccinimide ester

Sequence: Boc-Gly-OSu


Bachem additionally offers deuterated glycine: Fmoc-[D₂]Gly-OH B-4125. For N-methylglycine derivatives see Sarcosine.

Formula: C₁₁H₁₆N₂O₆
Storage Temperature: Freezer
MDL Number: MFCD00042761
CAS Number: 3392-07-2
EINECS: 222-230-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR