N-Boc-glycine (boc-gly-OH)

Supplier: BACHEM AMERICAS INC

4001229.0100 4001229.0500 4001229.1000
A-1730.0100BAEA 227.28 CAD
A-1730.0100BA A-1730.0500BA A-1730.1000BA
N-Boc-glycine (boc-gly-OH)
N-Boc-glycine (Boc-Gly-OH)

Sequence: Boc-Gly-OH


Bachem additionally offers deuterated glycine: Fmoc-[D₂]Gly-OH B-4125. For N-methylglycine derivatives see Sarcosine.

Formula: C₇H₁₃NO₄
Storage Temperature: Freezer
MDL Number: MFCD00002690
CAS Number: 4530-20-5
EINECS: 224-864-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR