Boc-Glu(OtBu)-OH
Supplier: BACHEM AMERICAS INC
4002938.0005
4002938.0025
A-1675.0005BAEA
185.3
CAD
A-1675.0005BA
A-1675.0025BA
Boc-Glu(OtBu)-OH
Boc-Glu(OtBu)-OH
Sequence: Boc-Glu(OtBu)-OH
The derivatives of Glu-NH₂ are assembled in the "Isoglutamine" subfamily.
Formula:
C₁₄H₂₅NO₆ MW: 303.355 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD00038257 CAS Number: 13726-84-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...