5-Benzyl-N-(tert-Butoxycarbonyl)-L-glutamate

Supplier: BACHEM AMERICAS INC

Synonyms: N-(tert-Butoxycarbonyl)-L-glutamic Acid 5-Benzyl Ester, N-Boc-L-glutamic Acid 5-Benzyl Ester, 5-Benzyl N-Boc-L-glutamate, Boc-Glu(OBzl)-OH

4000651.0025 4000651.0100
A-1640.0025BAEA 180.07 CAD
A-1640.0025BA A-1640.0100BA
5-Benzyl-N-(tert-Butoxycarbonyl)-L-glutamate
5-Benzyl-N-(tert-Butoxycarbonyl)-L-glutamate

Sequence: Boc-Glu(OBzl)-OH


The derivatives of Glu-NH₂ are assembled in the "Isoglutamine" subfamily.

Formula: C₁₇H₂₃NO₆
MW: 337.37 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00065569
CAS Number: 13574-13-5
EINECS: 237-007-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR