Boc-Lys(Fmoc)-OH

Supplier: BACHEM AMERICAS INC

4008005.0001 4008005.0005 4008005.0025
A-1610.0001BAEA 185.3 CAD
A-1610.0001BA A-1610.0005BA A-1610.0025BA
Boc-Lys(Fmoc)-OH
Boc-Lys(Fmoc)-OH

Sequence: Boc-Lys(Fmoc)-OH


For biotinylated lysine derivatives or chromophores and fluorophores coupled to Lys please see the subfamily "Nε-labeled Lysine".

Formula: C₂₆H₃₂N₂O₆
Storage Temperature: Ambient
MDL Number: MFCD00062032
CAS Number: 84624-27-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR