Boc-Cys(SEt)-OH·DCHA

Supplier: BACHEM AMERICAS INC

4000384.0001 4000384.0005
A-1600.0001BAEA 185.46 CAD
A-1600.0001BA A-1600.0005BA
Boc-Cys(SEt)-OH·DCHA
Boc-Cys(SEt)-OH·DCHA

Sequence: Boc-Cys(SEt)-OH · DCHA


The ethylthio group is cleaved by mercaptans.

Formula: C₁₀H₁₉NO₄S₂·C₁₂H₂₃N
Storage Temperature: Ambient
MDL Number: MFCD00236837
CAS Number: 25461-01-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR