Boc-Arg(Z)2-OSu

Supplier: BACHEM AMERICAS INC

Synonyms: Boc-Arg(Z)₂-OSu

4002887.0001 4002887.0005
A-1570.0001BAEA 370.51 CAD
A-1570.0001BA A-1570.0005BA
Boc-Arg(Z)2-OSu
Boc-Arg(Z)2-OSu

Sequence: Boc-Arg(Z)₂-OSu


Bachem additionally offers decarboxy-Arg: Agmatine · sulfate Q-1880.

Formula: C₃₁H₃₇N₅O₁₀
MW: 639.662 g/mol
Storage Temperature: Freezer
MDL Number: MFCD00236826
CAS Number: 76052-29-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR