Boc-3,4-dehydro-L-proline

Supplier: BACHEM AMERICAS INC

Synonyms: (S)-1-(tert-Butoxycarbonyl)-2,5-dihydro-1H-pyrrole-2-carboxylic acid, Boc-3,4-dehydro-Pro-OH

4004122.0001 4004122.0005
A-1550.0001BAEA 800.08 CAD
A-1550.0001BA A-1550.0005BA
Boc-3,4-dehydro-L-proline
Boc-3,4-dehydro-L-proline

Sequence: Boc-3,4-dehydro-Pro-OH


Dhp derivatives.


This dehydro-amino acid has been incorporated into biologically active peptides such as, e.g., oxytocin, bradykinin, Arg-vasopressin, or angiotensin I-converting enzyme inhibitors.

Formula: C₁₀H₁₅NO₄
MW: 213.233 g/mol
Storage Temperature: Freezer
MDL Number: MFCD00037896
CAS Number: 51154-06-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR