Boc-CIT-ONP

Supplier: BACHEM AMERICAS INC

4000081.0001 4000081.0005
A-1530.0001BAEA 232.35 CAD
A-1530.0001BA A-1530.0005BA
Boc-CIT-ONP
Boc-CIT-ONP

Sequence: Boc-Cit-ONp


Bachem also offers Fmoc-Cit-Wang resin, D-1840.


Synonym(s): Boc-Orn(carbamoyl)-ONp

Formula: C₁₇H₂₄N₄O₇
Storage Temperature: Freezer
MDL Number: MFCD00076998
CAS Number: 56612-88-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR