N-α-Boc-N-ε-(2-chloro-Z)-L-lysine

Supplier: BACHEM AMERICAS INC

4001418.0025 4001418.0100
A-1510.0025BAEA 276.63 CAD
A-1510.0025BA A-1510.0100BA
N-α-Boc-N-ε-(2-chloro-Z)-L-lysine
N-alpha-Boc-N-epsilon-(2-chloro-Z)-L-lysine

Sequence: Boc-Lys(2-chloro-Z)-OH


For biotinylated lysine derivatives or chromophores and fluorophores coupled to Lys please see the subfamily "Nε-labeled Lysine".


Synonym(s): Boc-Lys(2-Cl-Z)-OH

Formula: C₁₉H₂₇ClN₂O₆
Storage Temperature: Ambient
MDL Number: MFCD00038386
CAS Number: 54613-99-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR