Nɑ-(tert-Butoxycarbonyl)-Ndelta-carbobenzoxy-L-ornithine

Supplier: BACHEM AMERICAS INC

4005026.0025 4005026.0100
A-1490.0025BAEA 264.44 CAD
A-1490.0025BA A-1490.0100BA
Nɑ-(tert-Butoxycarbonyl)-Ndelta-carbobenzoxy-L-ornithine
Nalpha-(tert-Butoxycarbonyl)-Ndelta-carbobenzoxy-L-ornithine

Sequence: Boc-Orn(Z)-OH

Formula: C₁₈H₂₆N₂O₆
Storage Temperature: Ambient
MDL Number: MFCD00038259
CAS Number: 2480-93-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR