Boc-Lys(Z)-OSu
Supplier: BACHEM AMERICAS INC
4000178.0005
4000178.0025
A-1475.0005BAEA
237.27
CAD
A-1475.0005BA
A-1475.0025BA
Boc-Lys(Z)-OSu
Boc-Lys(Z)-OSu
Sequence: Boc-Lys(Z)-OSu
For biotinylated lysine derivatives or chromophores and fluorophores coupled to Lys please see the subfamily "Nε-labeled Lysine".
Formula:
C₂₃H₃₁N₃O₈ Storage Temperature: Freezer |
MDL Number:
MFCD00037915 CAS Number: 34404-36-9 EINECS: 251-999-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...