(S)-3-(4-(tert-Butoxy)phenyl)-2-((tert-butoxycarbonyl)amino)propanoic acid

Supplier: BACHEM AMERICAS INC

Synonyms: Boc-Tyr(tBu)-OH, N-Boc-O-tert-butyl-L-tyrosine

4007047.0005 4007047.0025
A-1450.0005BAEA 232.35 CAD
A-1450.0005BA A-1450.0025BA
(S)-3-(4-(tert-Butoxy)phenyl)-2-((tert-butoxycarbonyl)amino)propanoic acid
(S)-3-(4-(tert-Butoxy)phenyl)-2-((tert-butoxycarbonyl)amino)propanoic acid

Sequence: Boc-Tyr(tBu)-OH


Besides a large selection of tyrosine derivatives, Bachem offers derivatives of O-alkylated tyrosines, thyronine, phosphotyrosine and sulfotyrosine. Bachem additionally offers H-[¹⁵N]Tyr-OH, E-3245.

Formula: C₁₈H₂₇NO₅
Storage Temperature: Ambient
MDL Number: MFCD00065598
CAS Number: 47375-34-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR