N-Boc-O-tert-butyl-L-serine dicyclohexylammonium salt

Supplier: BACHEM AMERICAS INC

Synonyms: Boc-Ser(tBu)-OH · DCHA

4001527.0001 4001527.0005 4001527.0025
A-1430.0001BAEA 180.91 CAD
A-1430.0001BA A-1430.0005BA A-1430.0025BA
N-Boc-O-tert-butyl-L-serine dicyclohexylammonium salt
N-Boc-O-tert-butyl-L-serine dicyclohexylammonium salt

Storage temperature: +5 ± 3 °C

Formula: C₂₄H₄₆N₂O₅
MW: 442.641 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00065590
CAS Number: 18942-50-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR