N-(tert-Butoxycarbonyl)-O-benzyl-L-tyrosine

Supplier: BACHEM AMERICAS INC

Synonyms: N-Boc-O-benzyl-L-tyrosine, BOC-O-Benzyl-L-tyrosine, Boc-Tyr(Bzl)-OH

4000101.0025 4000101.0100
A-1390.0025BAEA 232.35 CAD
A-1390.0025BA A-1390.0100BA
N-(tert-Butoxycarbonyl)-O-benzyl-L-tyrosine
N-(tert-Butoxycarbonyl)-O-benzyl-L-tyrosine

Sequence: Boc-Tyr(Bzl)-OH


Besides a large selection of tyrosine derivatives, Bachem offers derivatives of O-alkylated tyrosines, thyronine, phosphotyrosine and sulfotyrosine. Bachem additionally offers H-[¹⁵N]Tyr-OH, E-3245.

Formula: C₂₁H₂₅NO₅
Storage Temperature: Ambient
MDL Number: MFCD00065597
CAS Number: 2130-96-3
EINECS: 218-349-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR