Boc-D-His(bom)-OH

Supplier: BACHEM AMERICAS INC

Synonyms: N-Boc-3-benzyloxymethyl-D-histidine, Boc-D-His(3-Bom)-OH

4011107.0001 4011107.0005
A-1375.0001BAEA 284.04 CAD
A-1375.0001BA A-1375.0005BA
Boc-D-His(bom)-OH
Boc-D-His(bom)-OH

Sequence: Boc-D-His(3-Bom)-OH


The atoms of the imidazole ring of histidine can be numbered in two different ways. Bachem as well as organic chemists designate the position of N-τ as 1 and the position of N-π as 3, whereas biochemists generally number N-π, the nitrogen atom adjacent to the side chain, as 1, and N-τ as 3.


Synonym(s): Boc-D-His(π-Bom)-OH

Formula: C₁₉H₂₅N₃O₅
MW: 375.425 g/mol
Storage Temperature: Freezer
MDL Number: MFCD00076940
CAS Number: 99310-01-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR