N-(tert-Butoxycarbonyl)-O-benzyl-L-threonine

Supplier: BACHEM AMERICAS INC

4000026.0025 4000026.0100
A-1355.0025BAEA 227.28 CAD
A-1355.0025BA A-1355.0100BA
N-(tert-Butoxycarbonyl)-O-benzyl-L-threonine
N-(tert-Butoxycarbonyl)-O-benzyl-L-threonine

Sequence: Boc-Thr(Bzl)-OH


Bachem also offers phosphothreonine derivatives.

Formula: C₁₆H₂₃NO₅
Storage Temperature: Ambient
MDL Number: MFCD00066062
CAS Number: 15260-10-3
EINECS: 239-304-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR