O-Benzyl-N-(tert-butoxycarbonyl)-D-serine

Supplier: BACHEM AMERICAS INC

4002168.0001 4002168.0025
A-1335.0001BAEA 175.66 CAD
A-1335.0001BA A-1335.0025BA
O-Benzyl-N-(tert-butoxycarbonyl)-D-serine
O-Benzyl-N-(tert-butoxycarbonyl)-D-serine

Sequence: Boc-D-Ser(Bzl)-OH


Bachem also offers derivatives of phosphoserine and sulfoserine.

Formula: C₁₅H₂₁NO₅
Storage Temperature: Ambient
MDL Number: MFCD00038248
CAS Number: 47173-80-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR