(S)-3-(Benzyloxy)-2-((tert-butoxycarbonyl)amino)propanoic acid

Supplier: BACHEM AMERICAS INC

Synonyms: Boc-Ser(Bzl)-OH, N-Boc-O-benzyl-L-serine

4001958.0025 4001958.0100
A-1330.0025BAEA 276.63 CAD
A-1330.0025BA A-1330.0100BA
(S)-3-(Benzyloxy)-2-((tert-butoxycarbonyl)amino)propanoic acid
(S)-3-(Benzyloxy)-2-((tert-butoxycarbonyl)amino)propanoic acid

Sequence: Boc-Ser(Bzl)-OH


Bachem also offers derivatives of phosphoserine and sulfoserine.

Formula: C₁₅H₂₁NO₅
MW: 295.3 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00066063
CAS Number: 23680-31-1
EINECS: 245-820-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR