Boc-L-His(Bzl)-OH mixture of isomers
Supplier: BACHEM AMERICAS INC
Synonyms:
Boc-His(Bzl)-OH, tele-Benzyl-Nalpha-(tert-butoxycarbonyl)-L-histidine
Sequence: Boc-His(Bzl)-OH
The atoms of the imidazole ring of histidine can be numbered in two different ways. Bachem as well as organic chemists designate the position of N-τ as 1 and the position of N-π as 3, whereas biochemists generally number N-π, the nitrogen atom adjacent to the side chain, as 1, and N-τ as 3.
Mix of τ-Bzl (mainly formed) and π-Bzl isomer. CAS Number (τ-isomer): 20898-44-6.
Formula:
C₁₈H₂₃N₃O₄ MW: 345.4 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00066093 CAS Number: 20898-44-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...