4-Benzyl-N-(tert-butoxycarbonyl)-L-aspartate

Supplier: BACHEM AMERICAS INC

Synonyms: BOC-L-Aspartic acid 4-benzylester, 4-Benzyl N-Boc-L-aspartate, Boc-Asp(Obzl)-OH, Boc-Asp(OBzl)-OH, N-Boc-L-aspartic Acid 4-Benzyl Ester, N-(tert-Butoxycarbonyl)-L-aspartic Acid 4-Benzyl Ester

4000620.0025 4000620.0100
A-1245.0025BAEA 180.91 CAD
A-1245.0025BA A-1245.0100BA
4-Benzyl-N-(tert-butoxycarbonyl)-L-aspartate
4-Benzyl-N-(tert-butoxycarbonyl)-L-aspartate

Sequence: Boc-Asp(OBzl)-OH


Asp-NH₂ is also called isoasparagine, so our Asp-NH₂ derivatives are assembled in the "Isoasparagine" subfamily.

Formula: C₁₆H₂₁NO₆
MW: 323.35 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00065564
CAS Number: 7536-58-5
EINECS: 231-406-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR