N-ɑ-Boc-L-asparagine

Supplier: BACHEM AMERICAS INC

Synonyms: Boc-L-Asparagine, Boc-Asn-OH

4000491.0025 4000491.0100
A-1215.0025BAEA 180.91 CAD
A-1215.0025BA A-1215.0100BA
N-ɑ-Boc-L-asparagine
N-alpha-Boc-L-asparagine

Sequence: Boc-Asn-OH


For isoasparagine derivatives please see the subfamily "Isoasparagine" filed under "Aspartic Acid".

Formula: C₉H₁₆N₂O₅
Storage Temperature: Ambient
MDL Number: MFCD00038152
CAS Number: 7536-55-2
EINECS: 231-405-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR