Boc-Arg-OH

Supplier: BACHEM AMERICAS INC

Synonyms: (S)-2-((tert-Butoxycarbonyl)amino)-5-guanidinopentanoic acid

4003812.0005 4003812.0025 4003812.0100
A-1200.0005BAEA 201.32 CAD
A-1200.0005BA A-1200.0025BA A-1200.0100BA
Boc-Arg-OH
Boc-Arg-OH

Sequence: Boc-Arg-OH


Bachem additionally offers decarboxy-Arg: Agmatine · sulfate Q-1880.

Formula: C₁₁H₂₂N₄O₄
MW: 274.321 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00042632
CAS Number: 13726-76-6
EINECS: 237-295-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR