Boc-6-Ahx-OSu

Supplier: BACHEM AMERICAS INC

Synonyms: 6-[(tert-Butoxycarbonyl)amino]hexanoic acid N-Succinimidyl ester, Boc-ε-aminocaproic acid-OSu, 6-(Boc-amino)hexanoic acid N-Succinimidyl ester, Boc-NH-C6-NHS, Boc-6-Ahx-OSu, 6-[(tert-Butoxycarbonyl)amino]hexanoic Acid N-Succinimidyl Ester, 2,5-Dioxopyrrolidin-1-yl 6-(tert-butoxycarbonylamino)hexanoate

4000016.0001 4000016.0005
A-1155.0001BAEA 183.09 CAD
A-1155.0001BA A-1155.0005BA
Boc-6-Ahx-OSu
Boc-6-Ahx-OSu

Sequence: Boc-ε-aminocaproic acid-OSu


Flexible spacer molecules of various length.
A flexible non-polar spacer moiety abbreviated as LC, εAca, or εAhx. Bachem also offers Fmoc-εAhx-Wang resin (D-1050).


Synonym(s): Boc-6-aminohexanoic acid-OSu#Boc-εAhx-OSu

Formula: C₁₅H₂₄N₂O₆
MW: 328.365 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00237379
CAS Number: 51513-80-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR