N-(tert-Butoxycarbonyl)-6-aminohexanoic acid

Supplier: BACHEM AMERICAS INC

Synonyms: 6-[(tert-Butoxycarbonyl)amino]hexanoic Acid, 6-(Boc-amino)hexanoic Acid, Boc-6-Ahx-OH, N-Boc-6-aminohexanoic Acid, Boc-ε-Acp-OH

4000005.0005 4000005.0025
A-1150.0005BAEA 211.7 CAD
A-1150.0005BA A-1150.0025BA
N-(tert-Butoxycarbonyl)-6-aminohexanoic acid
N-(tert-Butoxycarbonyl)-6-aminohexanoic acid

Sequence: Boc-ε-aminocaproic acid


Flexible spacer molecules of various length.
A flexible non-polar spacer moiety abbreviated as LC, εAca, or εAhx. Bachem also offers Fmoc-εAhx-Wang resin (D-1050).


Synonym(s): Boc-6-aminohexanoic acid#Boc-εAhx-OH

Formula: C₁₁H₂₁NO₄
MW: 231.29 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00037798
CAS Number: 6404-29-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR