N-(tert-Butoxycarbonyl)-4-aminobutyric acid

Supplier: BACHEM AMERICAS INC

4007074.0005 4007074.0025
A-1140.0005BAEA 185.3 CAD
A-1140.0005BA A-1140.0025BA
N-(tert-Butoxycarbonyl)-4-aminobutyric acid
N-(tert-Butoxycarbonyl)-4-aminobutyric acid

Sequence: Boc-γ-Abu-OH


Flexible spacer molecules of various length.


Synonym(s): Boc-GABA-OH

Formula: C₉H₁₇NO₄
Storage Temperature: Ambient
MDL Number: MFCD00037313
CAS Number: 57294-38-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR