Boc-Ala-Ala-Pro-OH
Supplier: BACHEM AMERICAS INC
4003849.0250
4003849.1000
A-1100.0250BAEA
387.21
CAD
A-1100.0250BA
A-1100.1000BA
Boc-Ala-Ala-Pro-OH
Proteins and Peptides
For Boc-dipeptides Boc-Xaa-Gly-OH and Boc-Xaa-Pro-OH, which can be used as building blocks during Boc-SPPS, please see the product family 'Boc-Dipeptide Building Blocks' in the 'Amino Acid Derivatives' section. This family also includes short protected peptide fragments suitable for solution synthesis and preparation of enzyme substrates and inhibitors.
Boc-AAP was used as building block for obtaining rhodamine-derived enzyme substrates.