Boc-D-Alanine

Supplier: BACHEM AMERICAS INC

Synonyms: N-Boc-D-alanine, Boc-D-Ala-OH

4000180.0025 4000180.0100
A-1065.0025BAEA 283.5 CAD
A-1065.0025BA A-1065.0100BA
Boc-D-Alanine
Boc-D-Alanine

Sequence: Boc-D-Ala-OH


Bachem additionally offers deuterated alanine: Fmoc-[D₄]Ala-OH (B-4130).

Formula: C₈H₁₅NO₄
MW: 189.2 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00063123
CAS Number: 7764-95-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR