N-Boc-L-alanine
Supplier: BACHEM AMERICAS INC
4000225.0100
4000225.0500
4000225.1000
A-1060.0100BAEA
238.9
CAD
A-1060.0100BA
A-1060.0500BA
A-1060.1000BA
N-Boc-L-alanine
N-Boc-L-alanine
Sequence: Boc-Ala-OH
Bachem additionally offers deuterated alanine: Fmoc-[D₄]Ala-OH (B-4130).
Formula:
C₈H₁₅NO₄ Storage Temperature: Ambient |
MDL Number:
MFCD00037225 CAS Number: 15761-38-3 EINECS: 239-847-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...