Nε-acetyl-Nα-boc-L-lysine

Supplier: BACHEM AMERICAS INC

Synonyms: Boc-Lys(Ac)-OH

4010020.0001 4010020.0005
A-1050.0001BAEA 185.3 CAD
A-1050.0001BA A-1050.0005BA
Nε-acetyl-Nα-boc-L-lysine
Nepsilon-acetyl-Nalpha-boc-L-lysine
Formula: C₁₃H₂₄N₂O₅
MW: 288.344 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00057791
CAS Number: 6404-26-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR