Boc-Tyr(Boc)-OH

Supplier: BACHEM AMERICAS INC

4000116.0005 4000116.0025
A-1040.0005BAEA 238.17 CAD
A-1040.0005BA A-1040.0025BA
Boc-Tyr(Boc)-OH
Boc-Tyr(Boc)-OH

Sequence: Boc-Tyr(Boc)-OH


Besides a large selection of tyrosine derivatives, Bachem offers derivatives of O-alkylated tyrosines, thyronine, phosphotyrosine and sulfotyrosine. Bachem additionally offers H-[¹⁵N]Tyr-OH, E-3245.

Formula: C₁₉H₂₇NO₇
Storage Temperature: Freezer
MDL Number: MFCD00038856
CAS Number: 20866-48-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR