Nɑ,N-ε-Bis(tert-butoxycarbonyl)-L-lysine dicyclohexylammonium salt

Supplier: BACHEM AMERICAS INC

Synonyms: Boc-Lys(Boc)-OH.DCHA, Boc-Lys(Boc)-OH·DCHA, Nα,Nepsilon-Di-Boc-L-lysine dicyclohexylammonium salt, Nɑ,N-ε-Di-Boc-L-lysine Dicyclohexylammonium Salt

4001224.0025 4001224.0100
A-1020.0025BAEA 244.24 CAD
A-1020.0025BA A-1020.0100BA
Nɑ,N-ε-Bis(tert-butoxycarbonyl)-L-lysine dicyclohexylammonium salt
Nalpha,N-epsilon-Bis(tert-butoxycarbonyl)-L-lysine dicyclohexylammonium salt

Sequence: Boc-Lys(Boc)-OH · DCHA


For biotinylated lysine derivatives or chromophores and fluorophores coupled to Lys please see the subfamily "Nε-labeled Lysine".


Building block for the synthesis of branched peptides and MAP matrices.

Formula: C₂₈H₅₃N₃O₆
Storage Temperature: Refrigerator
MDL Number: MFCD00038892
CAS Number: 15098-69-8
EINECS: 239-150-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR