Boc-His(1-boc)-OSu

Supplier: BACHEM AMERICAS INC

4010604.0001 4010604.0005 4010604.0025
A-1015.0001BAEA 180.91 CAD
A-1015.0001BA A-1015.0005BA A-1015.0025BA
Boc-His(1-boc)-OSu
Boc-His(1-boc)-OSu

Sequence: Boc-His(1-Boc)-OSu


The atoms of the imidazole ring of histidine can be numbered in two different ways. Bachem as well as organic chemists designate the position of N-τ as 1 and the position of N-π as 3, whereas biochemists generally number N-π, the nitrogen atom adjacent to the side chain, as 1, and N-τ as 3.


Synonym(s): Boc-His(τ-Boc)-OSu

Formula: C₂₀H₂₈N₄O₈
Storage Temperature: Freezer
MDL Number: MFCD00236852
CAS Number: 25616-02-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR