(Boc-Cys-OH)₂

Supplier: BACHEM AMERICAS INC

Synonyms: N,N'-di-Boc-L-cystine, AK-32714, (Boc-Cys-OH)2, Bis-Boc-L-cystine, (Boc-Cystine-OH)₂

4001680.0005 4001680.0025
A-1005.0005BAEA 183.09 CAD
A-1005.0005BA A-1005.0025BA
(Boc-Cys-OH)₂
(Boc-Cys-OH)₂

Sequence: (Boc-Cys-OH)₂


As the disulfide bond can be cleaved under mild conditions (e.g. with trialkylphosphines) cystine derivatives are highly economic educts for synthesizing Cys derivatives. The cystine derivatives of fluorophores (e.g. J-1175) or the pNA derivatives may be chosen as educts for synthesizing Cys substrates.


Tai et al. could obtain asymmetric cystine peptides from (Boc-Cys-OH)₂ by enzymatic catalysis with immobilized papain. Oxidation of Boc-cystine with hydrogen peroxide in the presence of Na wolframate yielded the sodium salt of Boc-cysteic acid. Boc cystine benzyl ester was used as educt for obtaining S-isoacyl dipeptide building blocks.

Formula: C₁₆H₂₈N₂O₈S₂
MW: 440.526 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00038250
CAS Number: 10389-65-8
EINECS: 233-852-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR