Human alpha-CGRP (8-37) Trifluoroacetate

Supplier: BACHEM AMERICAS INC

4013696.0001 4013696.0005
H-9895.0001BAEA 793.9 CAD
H-9895.0001BA H-9895.0005BA
Human alpha-CGRP (8-37) Trifluoroacetate
Proteins and Peptides

CGRP and calcitonin cross-react with each other at the levels of their distinct receptors. α-CGRP (8-37) has been described as a selective antagonist for CGRP receptors but not for calcitonin receptors. This fragment thus appears to be a valuable tool for receptor characterization studies.

Order Now

SPECIFICATIONS

Protein/peptide name alpha-CGRP (8-37) Trifluoroacetate
Environmentally Preferable
Protein synonyms Calcitonin Gene-Related Peptide I (8-37)|CGRP-I (8-37)
Species Human
Source
CAS No. 119911-68-1
MDL No. MFCD18782511
Sequence H-Val-Thr-His-Arg-Leu-Ala-Gly-Leu-Leu-Ser-Arg-Ser-Gly-Gly-Val-Val-Lys-Asn-Asn-Phe-Val-Pro-Thr-Asn-Val-Gly-Ser-Lys-Ala-Phe-NH₂
Molecular weight 3125.63
Molecular formula C₁₃₉H₂₃₀N₄₄O₃₈
Storage conditions –20 ± 5 °C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR