Mouse;Rat Amylin Trifluoroacetate
Supplier: BACHEM AMERICAS INC
4030201.0500 4030201.1000
H-9475.0500BAEA 814.24 CAD
H-9475.0500BA H-9475.1000BA
Mouse;Rat Amylin Trifluoroacetate
Proteins and Peptides
Rat amylin (rIAPP) lacks the fibril-forming capacity of human amylin (hIAPP). As a consequence, toxic effects have been reported for the human but not for the rat peptide amide. The sequence of H-9475 is identical to mouse IAPP.