(Sar⁹,Met(O₂)¹¹)-Substance P
Supplier: BACHEM AMERICAS INC
4013867.0001 4013867.0005
H-9410.0001BAEA 206.51 CAD
H-9410.0001BA H-9410.0005BA
(Sar⁹,Met(O₂)¹¹)-Substance P
Proteins and Peptides
This NK-1 receptor agonist exhibits the highest selectivity for NK-1 receptors in various bioassays. It evokes a concentration-dependent increase in mucus secretion and is much more potent than substance P (EC₅₀= 1.7 · 10-⁶M).