Kinetensin
Supplier: BACHEM AMERICAS INC
4016325.0005 4016325.0025
H-9350.0005BAEA 317.52 CAD
H-9350.0005BA H-9350.0025BA
Kinetensin
Proteins and Peptides
Kinetensin, a biologically active neurotensin-related peptide, has been obtained by pepsin treatment of human, bovine and dog plasma albumins. Its structure is similar to neurotensin and angiotensin I. Kinetensin increases vascular permeability when injected intradermally into rats and releases histamine from rat mast cells in vitro.