(N-Me-Phe⁷)-Neurokinin B Trifluoroacetate

Supplier: BACHEM AMERICAS INC

4016322.0001 4016322.0005
H-9280.0001BAEA 258.14 CAD
H-9280.0001BA H-9280.0005BA
(N-Me-Phe⁷)-Neurokinin B Trifluoroacetate
Proteins and Peptides

The MePhe-analog of NKB shows high selectivity for NKB receptors.

Order Now

SPECIFICATIONS

Protein/peptide name (N-Me-Phe⁷)-Neurokinin B Trifluoroacetate
Environmentally Preferable
Species
Source
CAS No. 110880-53-0
MDL No. MFCD18782503
Sequence H-Asp-Met-His-Asp-Phe-Phe-N-Me-Phe-Gly-Leu-Met-NH₂
Molecular weight 1272.51
Molecular formula C₆₀H₈₁N₁₃O₁₄S₂
Storage conditions –20 ± 5 °C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR