Bovine (D-Trp¹²,Tyr³⁴)-pTH (7-34) Amide Trifluoroacetate
Supplier: BACHEM AMERICAS INC
4016353.0500 4016353.1000
H-9115.0500BAEA 1135.93 CAD
H-9115.0500BA H-9115.1000BA
Bovine (D-Trp¹²,Tyr³⁴)-pTH (7-34) Amide Trifluoroacetate
Proteins and Peptides
See also the pTH2 receptor agonist TIP39 (H-4878).
The D-Trp¹² substitution leads to a competitive pTH antagonist with increased inhibitory properties in vitro.