Human;Mouse;Rat pTH-Related Protein (7-34) amide Trifluoroacetate
Supplier: BACHEM AMERICAS INC
4016931.0500 4016931.1000
H-9100.0500BAEA 529.25 CAD
H-9100.0500BA H-9100.1000BA
Human;Mouse;Rat pTH-Related Protein (7-34) amide Trifluoroacetate
Proteins and Peptides
pTHrP (7-34) amide is a potent antagonist of the effects of pTH-related protein (1-34) in vitro and in vivo.