omega-Conotoxin MVIIC
Supplier: BACHEM AMERICAS INC
4030522.0500 4030522.1000
H-8835.0500BAEA 1290.81 CAD
H-8835.0500BA H-8835.1000BA
omega-Conotoxin MVIIC
Proteins and Peptides
Like ω-conotoxin GVIA, this Conus magus peptide inhibits presynaptic Ca²⁺ channels. However, ω-conotoxin MVIIC clearly inhibits additional Ca²⁺ channel subtypes in the mammalian central nervous system. Thus, it should become a standard pharmacological reagent for investigating synaptic transmission in the mammalian nervous system.