Dolastatin 15
Supplier: BACHEM AMERICAS INC
4019226.0001 4019226.0005
H-8630.0001BAEA 407.1 CAD
H-8630.0001BA H-8630.0005BA
Dolastatin 15
Proteins and Peptides
Dolastatin 15 originally isolated from the sea hare Dolabella auricularia belongs to a family of antimitotic and antineoplastic depsipeptides inducing apoptosis in various malignant cell types. It was shown to inhibit the growth of the P388 lymphocytic leukemia cell line efficiently, with an ED₅₀ value of 2.4 ng/mL.