(Tyr⁵,D-Trp⁶·⁸·⁹,Lys-NH₂¹⁰)-Neurokinin A (4-10)
Supplier: BACHEM AMERICAS INC
4018827.0001 4018827.0005
H-8565.0001BAEA 258.14 CAD
H-8565.0001BA H-8565.0005BA
(Tyr⁵,D-Trp⁶·⁸·⁹,Lys-NH₂¹⁰)-Neurokinin A (4-10)
Proteins and Peptides
MEN10376 is a potent antagonist with high affinity and selectivity for the NK-2 receptor in vitro and in vivo.